Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tricosanoic acid | 2433-96-7 | 100.0% | 354.62 | 100 MG
SDP

Catalog No. 5000452643
Encompass_Preferred

Tricosanoic acid is an endogenous agonist of long-chain saturated fatty acids and free fatty acid receptor FFAR1, which can activate hair growth. It can improve cognitive function by regulating neuronal membrane fluidity, inhibiting neuroinflammatory response, participating in myelination, and neuronal energy metabolism. Levels are low in Alzheimer's disease (AD) models, while higher serum concentrations correlate with better cognitive performance. It can also be used as a biomarker for diseases related to cognitive decline.

  • Target: Free Fatty Acid Receptor
  • Research areas: Neurological disease, metabolic disease
  • Structural classification: Ketones, aldehydes, acids
  • Application fields: Metabolic disease, endogenous metabolite

Catalog No. 50-004-52643 Supplier Medchemexpress LLC Supplier No. HYW009081100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tricosanoic acid is an endogenous agonist of long-chain saturated fatty acids and free fatty acid receptor FFAR1, which can activate hair growth. It can improve cognitive function by regulating neuronal membrane fluidity, inhibiting neuroinflammatory response, participating in myelination, and neuronal energy metabolism. Levels are low in Alzheimer's disease (AD) models, while higher serum concentrations correlate with better cognitive performance. It can also be used as a biomarker for diseases related to cognitive decline.

  • Target: Free Fatty Acid Receptor
  • Research areas: Neurological disease, metabolic disease
  • Structural classification: Ketones, aldehydes, acids
  • Application fields: Metabolic disease, endogenous metabolite

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.