Learn More
Apexbio Technology LLC Tricosanoic acid 2433-96-7 200mg
SDP
Catalog No. 5000095975
Encompass_Preferred
Tricosanoic acid (CAS 2433-96-7) is a saturated long-chain fatty acid comprising 23 carbon atoms naturally present in plant oils animal fats and waxes While its precise molecular targets remain to be fully elucidated tricosanoic acid serves as a model compound in studies examining lipid metabolism membrane biophysics and the structural effects of very long-chain saturated fatty acids within biological membranes It is utilized in research exploring fatty acid incorporation lipid signaling pathways and the biophysical properties of lipid bilayers Its applications extend to investigations in metabolic disorders lipidomics and membrane dynamics
Catalog No. 50-000-95975 Supplier Apexbio Technology LLC Supplier No. A9968200
Tricosanoic acid (CAS 2433-96-7) is a saturated long-chain fatty acid comprising 23 carbon atoms naturally present in plant oils animal fats and waxes While its precise molecular targets remain to be fully elucidated tricosanoic acid serves as a model compound in studies examining lipid metabolism membrane biophysics and the structural effects of very long-chain saturated fatty acids within biological membranes It is utilized in research exploring fatty acid incorporation lipid signaling pathways and the biophysical properties of lipid bilayers Its applications extend to investigations in metabolic disorders lipidomics and membrane dynamics