Learn More
Medchemexpress LLC Tricosanoic acid | 2433-96-7 | 99.9% | 354.62 | 250 MG
SDP
Catalog No. 5000451574
Encompass_Preferred
Tricosanoic acid is an endogenous agonist of long-chain saturated fatty acids and free fatty acid receptor FFAR1. It can activate hair growth and improve cognitive function by regulating neuronal membrane fluidity, inhibiting neuroinflammatory response, and participating in myelination and neuronal energy metabolism. Low expression levels are observed in Alzheimer's disease models, with higher serum concentrations correlating to improved cognitive performance, making it a potential biomarker for cognitive decline.
  • Endogenous agonist of long-chain saturated fatty acids and free fatty acid receptor FFAR1
  • Activates hair growth
  • Improves cognitive function
  • Inhibits neuroinflammatory response
  • Participates in myelination and neuronal energy metabolism
  • Functions as a biomarker for cognitive decline
Catalog No. 50-004-51574 Supplier Medchemexpress LLC Supplier No. 250MGHYW009081
Tricosanoic acid is an endogenous agonist of long-chain saturated fatty acids and free fatty acid receptor FFAR1. It can activate hair growth and improve cognitive function by regulating neuronal membrane fluidity, inhibiting neuroinflammatory response, and participating in myelination and neuronal energy metabolism. Low expression levels are observed in Alzheimer's disease models, with higher serum concentrations correlating to improved cognitive performance, making it a potential biomarker for cognitive decline.
  • Endogenous agonist of long-chain saturated fatty acids and free fatty acid receptor FFAR1
  • Activates hair growth
  • Improves cognitive function
  • Inhibits neuroinflammatory response
  • Participates in myelination and neuronal energy metabolism
  • Functions as a biomarker for cognitive decline