Learn More
Cayman Chemical Tricosanoic Acid ethyl ester
SDP
Catalog No. 502889962
Encompass_Preferred
A saturated fatty acid ethyl ester; can be used as a standard for analysis of lipid mixtures
Catalog No. 50-288-9962 Supplier Cayman Chemical Supplier No. 1000687410 MG
Add to Cart
Add to Cart
A saturated fatty acid ethyl ester; can be used as a standard for analysis of lipid mixtures