Learn More
Cayman Chemical Tricosanoic Acid methyl ester, Purity: ≥98%, CAS: 2433-97-8, 500 mg
SDP
Catalog No. 502934539
Encompass_Preferred
A fatty acid methyl ester; has been found in peanut butter and in latent fingerprint residue samples
Catalog No. 50-293-4539 Supplier Cayman Chemical Supplier No. 26868500 MG
A fatty acid methyl ester; has been found in peanut butter and in latent fingerprint residue samples