Learn More
Cayman Chemical Tricosanoic Acid, Purity: ≥98%, CAS: 2433-96-7, 100 mg
SDP
Catalog No. 502934693
Encompass_Preferred
A naturally occurring saturated fatty acid; inhibits lysis of human erythrocytes induced by S. aureus delta toxin in a concentration-dependent manner; decreased in the sebum of patients with papulopustular rosacea; increased in malignant, but not adjacent nonmalignant, prostate tissue in patients with prostate cancer; has been used as an internal standard for the quantification of fatty acids in chicken egg yolk
Catalog No. 50-293-4693 Supplier Cayman Chemical Supplier No. 26867100 MG
A naturally occurring saturated fatty acid; inhibits lysis of human erythrocytes induced by S. aureus delta toxin in a concentration-dependent manner; decreased in the sebum of patients with papulopustular rosacea; increased in malignant, but not adjacent nonmalignant, prostate tissue in patients with prostate cancer; has been used as an internal standard for the quantification of fatty acids in chicken egg yolk