Learn More
Cayman Chemical Tricosanoyl Ethanolamide, Purity: ≥98%, CAS: 171022-15-4, 50 mg
SDP
Catalog No. 502893087
Encompass_Preferred
A member of the family of fatty N-acyl ethanolamines collectively called endocannabinoids
Catalog No. 50-289-3087 Supplier Cayman Chemical Supplier No. 900174350 MG
A member of the family of fatty N-acyl ethanolamines collectively called endocannabinoids