Learn More
Electron Microscopy Sciences Tricresyl Phosphate 225 ML
SDP
Supplier:  Electron Microscopy Sciences 22150
Encompass_Preferred
(Tritolyl Phosphate)
CAS #1330-78-5 (CH₃C₆H₄O)₃PO F.W. 368.36
A mixture of isomeric Tritolyl Phosphates, usually excluding the very toxic group ortho-isomer. Used to prepare benzoyl peroxide paste, which acts as a catalyst for water miscible resins. (Tritolyl Phosphate)
Catalog No. 50-295-14