Learn More
Medchemexpress LLC HY-B0848 100mg Medchemexpress, Tricyclazole CAS:41814-78-2 Purity:>98%
SDP
Catalog No. 501970127
Encompass_Preferred
Medchemexpress, HY-B0848 100mg Tricyclazole CAS:41814-78-2 Tricyclazole is a pentaketide-derived melanin biosynthesis inhibitor and a unique fungicide for control of Pyricularia oryzae on rice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0127 Supplier Medchemexpress LLC Supplier No. HYB0848100MG
Medchemexpress, HY-B0848 100mg Tricyclazole CAS:41814-78-2 Tricyclazole is a pentaketide-derived melanin biosynthesis inhibitor and a unique fungicide for control of Pyricularia oryzae on rice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.