Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tricyclazole, >95% min. , Ultra Scientific

Catalog No. USPST1840
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-PST-1840 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-PST-1840 Supplier Agilent Technologies Supplier No. PST1840
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 41814-78-2
Molecular Formula C9H7N3S
Molecular Weight (g/mol) 189.236
InChI Key DQJCHOQLCLEDLL-UHFFFAOYSA-N
Synonym 5-methyl-1,2,4-triazolo[3,4-b]benzothiazole
PubChem CID 39040
ChEBI CHEBI:81783
IUPAC Name 8-methyl-[1,2,4]triazolo[3,4-b][1,3]benzothiazole
SMILES CC1=C2C(=CC=C1)SC3=NN=CN23

Specifications

CAS Max % 100.0
Assay Percent Range >95 % min.
Solvent or Matrix Neat
Quantity 10 mg
Chemical Name or Material Tricyclazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.