Learn More
Medchemexpress LLC Tricyclo[3.3.1.13,7]decan-1-ol | 768-95-6 | MFCD00074729 | 99.6% | 152.24 | C10H16O | 25g
SDP
Catalog No. 5000268926
Encompass_Preferred
1-Adamantanol is a cyclic molecule with a hydroxyl group that can be used as an intermediate in the synthesis of adamantane 1-Adamantanol can be oxidized to 1 3-adamantanediol by the Streptomyces SA8 oxidation system[1]
Catalog No. 50-002-68926 Supplier Medchemexpress LLC Supplier No. HYY053125G
1-Adamantanol is a cyclic molecule with a hydroxyl group that can be used as an intermediate in the synthesis of adamantane 1-Adamantanol can be oxidized to 1 3-adamantanediol by the Streptomyces SA8 oxidation system[1]