Learn More
Strem, An Ascensus Company CAS# 58656-04-5. 1g. Tricyclohexylphosphonium tetrafluoroborate, 99%. MFCD03840580
SDP
Catalog No. 5090110393
Encompass
CAS# 58656-04-5. 1g. Tricyclohexylphosphonium tetrafluoroborate, 99%. MFCD03840580. Molecular Weight: 368.24. Molecular Formula: [(C6H11)3PH]+BF4ae. Color/form: white pwdr. Strem# 15-6160. http://www.strem.com/catalog/v/15-6160/
Catalog No. 50-901-10393 Supplier Strem, An Ascensus Company Supplier No. 1561601G
CAS# 58656-04-5. 1g. Tricyclohexylphosphonium tetrafluoroborate, 99%. MFCD03840580. Molecular Weight: 368.24. Molecular Formula: [(C6H11)3PH]+BF4ae. Color/form: white pwdr. Strem# 15-6160. http://www.strem.com/catalog/v/15-6160/