Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tricyclohexylphosphonium tetrafluoroborate, 99%

Catalog No. AAH2742814
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2742814 25 g
AAH2742803 1 g
AAH2742806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2742814 Supplier Thermo Scientific Chemicals Supplier No. H2742814
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 58656-04-5 | C18H34BF4P | 368.25 g/mol

Tricyclohexylphosphonium tetrafluoroborate is used with ruthenium (1,5-cyclooctadiene)ruthenium dimer to catalyze the dehydrogenative coupling of alcohols and amines to form amide bonds. It is also used in suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tricyclohexylphosphonium tetrafluoroborate is used with ruthenium (1,5-cyclooctadiene)ruthenium dimer to catalyze the dehydrogenative coupling of alcohols and amines to form amide bonds. It is also used in suzuki reaction.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58656-04-5
Molecular Formula C18H34BF4P
Molecular Weight (g/mol) 368.25
MDL Number MFCD03840580
InChI Key MYSMMEUXKHJYKH-UHFFFAOYSA-O
Synonym tricyclohexylphosphonium tetrafluoroborate, tricyclohexylphosphine tetrafluoroborate, tricyclohexylphosphine tetrafluroborate, tricyclohexylphosphoniumtetrafluoroborate, pcy3.hbf4, ksc269e4n, tricyclohexylphosphine tetrofluroborate, tricyclohexyl phosphonium tetrafluoroborate, tetrafluoroboranuide tricyclohexylphosphanium, tricyclohexyl-phosphonium tetrafluoro borate
PubChem CID 11710396
IUPAC Name tricyclohexylphosphanium;tetrafluoroborate
SMILES F[B-](F)(F)F.C1CCC(CC1)[PH+](C1CCCCC1)C1CCCCC1

Specifications

Melting Point 166°C to 170°C
Linear Formula (C6H11)3PH€BF4
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 368.25
Percent Purity 99%
Chemical Name or Material Tricyclohexylphosphonium tetrafluoroborate
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.