Learn More
Cayman Chemical Tridecanoic Acid methyl ester, Purity: ≥98%, CAS: 1731-88-0, 25 g
SDP
Catalog No. 502934955
Encompass_Preferred
An esterified form of tridecanoic acid; a component of biodiesel generated from SCW-SCM separation of sewage sludge
Catalog No. 50-293-4955 Supplier Cayman Chemical Supplier No. 2672125 G
An esterified form of tridecanoic acid; a component of biodiesel generated from SCW-SCM separation of sewage sludge