Learn More
Medchemexpress LLC Tridecane | 629-50-5 | 184.36 | C13H28 | 100 G
SDP
Catalog No. 5000208355
Encompass_Preferred
Tridecane is a straight-chain aliphatic hydrocarbon used as an analytical standard and reference for instrument calibration in analytical chemistry.
  • used as an analytical standard for GC/MS and instrument calibration.
  • straight-chain alkane with formula C13H28 and molecular weight 184.36.
  • listed CAS number 629-50-5 for unambiguous identification.
  • supplied in small pack sizes for analytical applications (manufacturer lists up to 5 g; supplier row indicates 100 g).
Catalog No. 50-002-08355 Supplier Medchemexpress LLC Supplier No. HYW088037100G
Tridecane is a straight-chain aliphatic hydrocarbon used as an analytical standard and reference for instrument calibration in analytical chemistry.
  • used as an analytical standard for GC/MS and instrument calibration.
  • straight-chain alkane with formula C13H28 and molecular weight 184.36.
  • listed CAS number 629-50-5 for unambiguous identification.
  • supplied in small pack sizes for analytical applications (manufacturer lists up to 5 g; supplier row indicates 100 g).