Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tridecane | 629-50-5 | 99.5% | 184.36 g/mol | C13H28 | 25 G
SDP

Catalog No. 5000208743
Encompass_Preferred

Tridecane is an analytical standard of the C13 aliphatic hydrocarbon tridecane, supplied for research and analytical applications such as chromatographic reference and calibration.

  • Analytical standard for chromatographic reference and calibration.
  • High purity: 99.51% (GC).
  • Low water content: 0.02% (KF).
  • Chemical formula: C13H28.
  • Molecular weight: 184.36 g/mol.

Catalog No. 50-002-08743 Supplier Medchemexpress LLC Supplier No. 25GHYW088037
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tridecane is an analytical standard of the C13 aliphatic hydrocarbon tridecane, supplied for research and analytical applications such as chromatographic reference and calibration.

  • Analytical standard for chromatographic reference and calibration.
  • High purity: 99.51% (GC).
  • Low water content: 0.02% (KF).
  • Chemical formula: C13H28.
  • Molecular weight: 184.36 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.