Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tridecane | 629-50-5 | MFCD00008979 | 184.36 | C13H28
SDP

Catalog No. 5000208435
Encompass_Preferred

Tridecane is a straight-chain aliphatic hydrocarbon (C13H28) used as an analytical standard and reference material for laboratory and analytical applications. The material is supplied in solid or neat form with documentation (certificate of analysis) for purity and storage; stock solutions (for example, 10 mM) can be prepared from the material using the provided preparation guidance.

  • Aliphatic hydrocarbon, chemical formula C13H28.
  • Molecular weight 184.36 g/mol.
  • CAS number 629-50-5.
  • Certificate of Analysis available for purity and storage instructions.
  • Preparation guidance provided for making stock solutions (example: 10 mM).

Catalog No. 50-002-08435 Supplier Medchemexpress LLC Supplier No. HYW08803710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tridecane is a straight-chain aliphatic hydrocarbon (C13H28) used as an analytical standard and reference material for laboratory and analytical applications. The material is supplied in solid or neat form with documentation (certificate of analysis) for purity and storage; stock solutions (for example, 10 mM) can be prepared from the material using the provided preparation guidance.

  • Aliphatic hydrocarbon, chemical formula C13H28.
  • Molecular weight 184.36 g/mol.
  • CAS number 629-50-5.
  • Certificate of Analysis available for purity and storage instructions.
  • Preparation guidance provided for making stock solutions (example: 10 mM).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.