Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tridecane | 629-50-5 | MFCD00008979 | 99.5% | 184.36 g/mol | C13H28 | 50 G
SDP

Catalog No. 5000208372
Encompass_Preferred

Tridecane is a straight-chain aliphatic hydrocarbon (C13H28) supplied as a liquid analytical standard for research and reference. It is commonly found in essential oils and may be used in hydrocarbon analysis and method development.

  • C13 aliphatic hydrocarbon
  • Purity 99.51%
  • Molecular weight 184.36 g/mol
  • Available as liquid in a 50 G pack size
  • Storage: -20°C for long-term storage, 4°C for shorter-term storage

Catalog No. 50-002-08372 Supplier Medchemexpress LLC Supplier No. HYW08803750G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tridecane is a straight-chain aliphatic hydrocarbon (C13H28) supplied as a liquid analytical standard for research and reference. It is commonly found in essential oils and may be used in hydrocarbon analysis and method development.

  • C13 aliphatic hydrocarbon
  • Purity 99.51%
  • Molecular weight 184.36 g/mol
  • Available as liquid in a 50 G pack size
  • Storage: -20°C for long-term storage, 4°C for shorter-term storage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.