Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tridecane, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014942 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4942 5 mL
11-101-4941 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-4942 Supplier MilliporeSigma Supelco Supplier No. 914905ML
Add to Cart
Add to Cart
Petroleum
TRUSTED_SUSTAINABILITY

Specifications

CAS 629-50-5
Vapor Pressure 1 mmHg (59.4°C)
Vapor Density 6.4 (vs. air)
Density 0.756 g/mL (at 25°C)
Flash Point 79°C
Physical Form Neat
Linear Formula CH3(CH2)11 CH3
MDL Number MFCD00008979
Melting Point -6°C to -4°C
Boiling Point 110°C to 112°C (12 mmHg); 234°C
Type Analytical Standard
Quantity 5 mL
Formula Weight 184.36
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.