Learn More
Medchemexpress LLC Tridecane-d28 | 121578-12-9 | MFCD00145015 | 98.1% | 212.53 g/mol | C13D28 | 10 MG
SDP
Catalog No. 5000208427
Encompass_Preferred
Tridecane-d28 is the deuterium-labeled form of tridecane used as an isotopic reference standard for analytical and research applications. It is fully deuterated (C13D28), with high isotopic enrichment and is supplied in small pack sizes suitable for analytical use.
  • fully deuterated alkane, useful as an internal standard for mass spectrometry
  • reported purity about 98.05%
  • molecular weight 212.53 g/mol and formula C13D28
  • available in small research pack sizes for analytical workflows
  • volatility similar to tridecane, suitable for GC applications
Catalog No. 50-002-08427 Supplier Medchemexpress LLC Supplier No. HYW088037S10MG
Tridecane-d28 is the deuterium-labeled form of tridecane used as an isotopic reference standard for analytical and research applications. It is fully deuterated (C13D28), with high isotopic enrichment and is supplied in small pack sizes suitable for analytical use.
  • fully deuterated alkane, useful as an internal standard for mass spectrometry
  • reported purity about 98.05%
  • molecular weight 212.53 g/mol and formula C13D28
  • available in small research pack sizes for analytical workflows
  • volatility similar to tridecane, suitable for GC applications