Learn More
Medchemexpress LLC Tridecanedioic acid | 505-52-2 | 99.9% | 100 G
SDP
Catalog No. 5000338921
Encompass_Preferred
Tridecanedioic acid is an endogenous metabolite related to the metabolic regulation of non-alcoholic fatty liver disease (NASH). It plays a significant role in the intestinal microbiota-host metabolism interaction network and accumulates in tolerant cabbage-type rapeseed varieties, contributing to defense against *Plutella xylostella* infection. This makes it suitable as a biomarker for plant insect resistance or a diagnostic marker for metabolic diseases.
  • Endogenous metabolite
  • Related to metabolic regulation of non-alcoholic fatty liver disease (NASH)
  • May be a key molecule in intestinal microbiota-host metabolism interaction
  • Accumulates in tolerant cabbage-type rapeseed varieties
  • Plays a role in defense against *Plutella xylostella* infection
  • Can serve as a biomarker for plant insect resistance
  • Can serve as a diagnostic marker for metabolic diseases
Catalog No. 50-003-38921 Supplier Medchemexpress LLC Supplier No. HY128421100G
Tridecanedioic acid is an endogenous metabolite related to the metabolic regulation of non-alcoholic fatty liver disease (NASH). It plays a significant role in the intestinal microbiota-host metabolism interaction network and accumulates in tolerant cabbage-type rapeseed varieties, contributing to defense against *Plutella xylostella* infection. This makes it suitable as a biomarker for plant insect resistance or a diagnostic marker for metabolic diseases.
  • Endogenous metabolite
  • Related to metabolic regulation of non-alcoholic fatty liver disease (NASH)
  • May be a key molecule in intestinal microbiota-host metabolism interaction
  • Accumulates in tolerant cabbage-type rapeseed varieties
  • Plays a role in defense against *Plutella xylostella* infection
  • Can serve as a biomarker for plant insect resistance
  • Can serve as a diagnostic marker for metabolic diseases