Learn More
Medchemexpress LLC Tridecanedioic acid | 505-52-2 | 99.9% | 25 G
SDP
Catalog No. 5000341478
Encompass_Preferred
Tridecanedioic acid, also known as Brassylic Acid, is an endogenous metabolite. It is associated with the metabolic regulation of non-alcoholic fatty liver disease (NASH) and may serve as a crucial node molecule within the interaction network between intestinal microbiota and host metabolism.
  • Significantly accumulates in tolerant cabbage-type rapeseed varieties
  • Plays an important role in defense against small cabbage moth infection
  • Can be used as a biomarker for plant insect resistance
  • Can be used as a diagnostic marker for metabolic diseases
Catalog No. 50-003-41478 Supplier Medchemexpress LLC Supplier No. 25GHY128421
Add to Cart
Add to Cart
Tridecanedioic acid, also known as Brassylic Acid, is an endogenous metabolite. It is associated with the metabolic regulation of non-alcoholic fatty liver disease (NASH) and may serve as a crucial node molecule within the interaction network between intestinal microbiota and host metabolism.
  • Significantly accumulates in tolerant cabbage-type rapeseed varieties
  • Plays an important role in defense against small cabbage moth infection
  • Can be used as a biomarker for plant insect resistance
  • Can be used as a diagnostic marker for metabolic diseases