Learn More
Medchemexpress LLC Tridecanoic acid-d2 | 64118-44-1 | 5mg
SDP
Catalog No. 5000265944
Encompass_Preferred
Tridecanoic acid-d2 is the deuterium labeled Tridecanoic acid Tridecanoic acid (N-Tridecanoic acid) a 13-carbon medium-chain saturated fatty acid can serve as an antipersister and antibiofilm agent that may be applied to research bacterial infections Tridecanoic acid inhibits Escherichia coli persistence and biofilm formation[1]
Catalog No. 50-002-65944 Supplier Medchemexpress LLC Supplier No. HYY1718S5MG
Tridecanoic acid-d2 is the deuterium labeled Tridecanoic acid Tridecanoic acid (N-Tridecanoic acid) a 13-carbon medium-chain saturated fatty acid can serve as an antipersister and antibiofilm agent that may be applied to research bacterial infections Tridecanoic acid inhibits Escherichia coli persistence and biofilm formation[1]