Learn More
Medchemexpress LLC Tridecanoic acid-d25 | 202529-03-1 | MFCD00053327 | 25mg
SDP
Catalog No. 5000265946
Encompass_Preferred
Tridecanoic acid-d25 is the deuterium labeled Tridecanoic acid Tridecanoic acid (N-Tridecanoic acid) a 13-carbon medium-chain saturated fatty acid can serve as an antipersister and antibiofilm agent that may be applied to research bacterial infections Tridecanoic acid inhibits Escherichia coli persistence and biofilm formation[1]
Catalog No. 50-002-65946 Supplier Medchemexpress LLC Supplier No. HYY1718S125MG
Tridecanoic acid-d25 is the deuterium labeled Tridecanoic acid Tridecanoic acid (N-Tridecanoic acid) a 13-carbon medium-chain saturated fatty acid can serve as an antipersister and antibiofilm agent that may be applied to research bacterial infections Tridecanoic acid inhibits Escherichia coli persistence and biofilm formation[1]