Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tridecanol (mixture of isomers), TCI America™
SDP

Catalog No. T041425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T041425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T041425ML Supplier TCI America Supplier No. T041425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26248-42-0
Molecular Formula C13H28O
Molecular Weight (g/mol) 200.37
MDL Number MFCD00004756
InChI Key XFRVVPUIAFSTFO-UHFFFAOYSA-N
Synonym Tridecyl Alcohol
PubChem CID 8207
ChEBI CHEBI:34123
IUPAC Name tridecan-1-ol
SMILES CCCCCCCCCCCCCO

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 200.37
Physical Form Liquid
Chemical Name or Material Tridecanol (mixture of isomers)
EINECSNumber (2)-0217&(2)-3704
RTECSNumber YD4150000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.