Learn More
Cayman Chemical Tridecanoic Acid, Purity: ≥95%, CAS: 638-53-9, 25 g
SDP
Catalog No. 502983554
Encompass_Preferred
A 13-carbon saturated fatty acid found in dairy products and also as a product of anaerobic biodegradation of n-hexadecane; acts as a substrate of phospholipase A2
Catalog No. 50-298-3554 Supplier Cayman Chemical Supplier No. 1972525 G
A 13-carbon saturated fatty acid found in dairy products and also as a product of anaerobic biodegradation of n-hexadecane; acts as a substrate of phospholipase A2