Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tridecyl Acetate 98.0+%, TCI America™
SDP

Catalog No. A145225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A145225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A145225G Supplier TCI America Supplier No. A145225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1072-33-9
Molecular Formula C15H30O2
Molecular Weight (g/mol) 242.403
MDL Number MFCD00056197
InChI Key ZDRNMODJXFOYMN-UHFFFAOYSA-N
Synonym Acetic Acid Tridecyl Ester
PubChem CID 14071
IUPAC Name tridecyl acetate
SMILES CCCCCCCCCCCCCOC(=O)C

Specifications

Color Colorless
Boiling Point 143°C
Quantity 25 g
Formula Weight 242.40
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tridecyl Acetate
EINECSNumber (2)-0735
RTECSNumber YD4300000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.