Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tridecylbenzene, Thermo Scientific™

Catalog No. AC151030250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC151030250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC151030250 Supplier Thermo Scientific Chemicals Supplier No. 151030250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-02-4
Molecular Formula C19H32
Molecular Weight (g/mol) 260.465
InChI Key MCVUKOYZUCWLQQ-UHFFFAOYSA-N
Synonym 1-phenyltridecane, benzene, tridecyl, n-tridecylbenzene, tridane, tridecane, 1-phenyl, benzene, c10-16-alkyl derivs., detergent alkylate no. 5, unii-e6k14656yo, c8-c16 alkylbenzene, benzene, mono-c12-14-alkyl derivs.
PubChem CID 31238
IUPAC Name tridecylbenzene
SMILES CCCCCCCCCCCCCC1=CC=CC=C1

Specifications

Density 0.85
Linear Formula C6H5(CH2)12CH3
Quantity 25 g
Merck Index 15, 9824
Formula Weight 260.45
Chemical Name or Material Tridecylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.