Learn More
Glass Expansion Trident CT Mixing Chamber
SDP
Catalog No. 502703092
Encompass_Preferred
Trident CT Mixing Chamber. Replacement Trident CT mixing chamber for Glass Expansion’s Trident CT Internal Standard (IS) Kit (P/Ns NC2046960, 60-703-1179). Not for use with HF. For solutions containing HF use Trident IS Kit for HF P/N NC2046964, 60-808-1150.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3092 Supplier Glass Expansion Supplier No. 607031119
Trident CT Mixing Chamber. Replacement Trident CT mixing chamber for Glass Expansion’s Trident CT Internal Standard (IS) Kit (P/Ns NC2046960, 60-703-1179). Not for use with HF. For solutions containing HF use Trident IS Kit for HF P/N NC2046964, 60-808-1150.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001