Learn More
Genetex Inc Trident Endosome Isolation Kit
SDP
Catalog No. NC1535070
Encompass
The Endosome Isolation and Cell Fractionation Kit provides a spin-column based novel endosome isolation technology that is rapid, simple and requires smaller number of cultured cells or milligram amounts of tissues. This kit can precipitate and significantly enrich early endosomes from cultured cells or tissues.
Catalog No. NC1535070 Supplier Genetex Inc Supplier No. GTX35192
The Endosome Isolation and Cell Fractionation Kit provides a spin-column based novel endosome isolation technology that is rapid, simple and requires smaller number of cultured cells or milligram amounts of tissues. This kit can precipitate and significantly enrich early endosomes from cultured cells or tissues.