Learn More
Medchemexpress LLC Trideuteriomethyl N-[(E)-quinoxalin-2-ylmethylideneamino]carbamate | 1448350-02-4 | 99.8% | C11H10N4O2 | 10MG
SDP
Catalog No. 5000259221
Encompass_Preferred
Desoxycarbadox-d3 is a deuterium-labeled analogue of desoxycarbadox, the primary metabolite of the veterinary antibiotic carbadox. Supplied as a high-purity isotopic standard, it is intended for use as an internal standard or tracer in quantitative mass spectrometry, method development, and pharmacokinetic studies.
  • Deuterium-labeled internal standard for LC-MS quantitation.
  • Contains three deuterium atoms for a clear mass shift.
  • High purity (99.8%) for reliable analytical results.
  • Stable white solid suitable for storage and handling.
  • Suitable for tracer and pharmacokinetic studies in biological matrices.
Catalog No. 50-002-59221 Supplier Medchemexpress LLC Supplier No. HY136428S10MG
Desoxycarbadox-d3 is a deuterium-labeled analogue of desoxycarbadox, the primary metabolite of the veterinary antibiotic carbadox. Supplied as a high-purity isotopic standard, it is intended for use as an internal standard or tracer in quantitative mass spectrometry, method development, and pharmacokinetic studies.
  • Deuterium-labeled internal standard for LC-MS quantitation.
  • Contains three deuterium atoms for a clear mass shift.
  • High purity (99.8%) for reliable analytical results.
  • Stable white solid suitable for storage and handling.
  • Suitable for tracer and pharmacokinetic studies in biological matrices.