Learn More
New England Biolabs, Inc. TriDye™ 1 kb DNA Ladder – 1.25 ml
SDP
Catalog No. 50811783
Encompass
TriDye 1 kb DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing 3 dyes which serve as visual aids to monitor the progress of migration during agarose gel electrophoresis. The DNA Ladder consists of proprietary plasmids which are digested to completion with appropriate restriction enzymes to yield 10 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The digested DNA includes fragments ranging from 0.5-10.0 kilobases. The 3.0 kb fragment has increased intensity to serve as a reference band.
Catalog No. 50-811-783 Supplier New England Biolabs, Inc. Supplier No. N3272S
TriDye 1 kb DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing 3 dyes which serve as visual aids to monitor the progress of migration during agarose gel electrophoresis. The DNA Ladder consists of proprietary plasmids which are digested to completion with appropriate restriction enzymes to yield 10 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The digested DNA includes fragments ranging from 0.5-10.0 kilobases. The 3.0 kb fragment has increased intensity to serve as a reference band.