Learn More
New England Biolabs, Inc. TriDye™ 100 bp DNA Ladder – 1.25 ml
SDP
Catalog No. 50811782
Encompass
TriDye 100 bp DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing 3 dyes which serve as visual aids to monitor the progress of migration during agarose gel electrophoresis. The DNA ladder consists of proprietary plasmids are digested to completion with appropriate restriction enzymes to yield 12 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The digested DNA includes fragments ranging from 100-1,517 base pairs. The 500 and 1,000 base pair bands have increased intensity to serve as reference points.
Catalog No. 50-811-782 Supplier New England Biolabs, Inc. Supplier No. N3271S
TriDye 100 bp DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing 3 dyes which serve as visual aids to monitor the progress of migration during agarose gel electrophoresis. The DNA ladder consists of proprietary plasmids are digested to completion with appropriate restriction enzymes to yield 12 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The digested DNA includes fragments ranging from 100-1,517 base pairs. The 500 and 1,000 base pair bands have increased intensity to serve as reference points.