Learn More
New England Biolabs, Inc. TriDye™ Ultra Low Range DNA Ladder – 1.25 ml
SDP
Catalog No. 501959410
Encompass
TriDye Ultra Low Range DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing three tracking dyes which serve as visual aids to monitor the progress of migration during native agarose and polyacrylamide gel electrophoresis. The DNA ladder contains 13 bands ranging from 10 bp to 700 bp, most of which are generated from the digestion of a proprietary plasmid with restriction enzymes. The 50 bp and 300 bp bands have increased intensity to serve as reference bands. The ladder is suitable for use with for use with both native polyacrylamide and agarose gel electrophoresis.
Catalog No. 50-195-9410 Supplier New England Biolabs, Inc. Supplier No. N0558S
TriDye Ultra Low Range DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing three tracking dyes which serve as visual aids to monitor the progress of migration during native agarose and polyacrylamide gel electrophoresis. The DNA ladder contains 13 bands ranging from 10 bp to 700 bp, most of which are generated from the digestion of a proprietary plasmid with restriction enzymes. The 50 bp and 300 bp bands have increased intensity to serve as reference bands. The ladder is suitable for use with for use with both native polyacrylamide and agarose gel electrophoresis.