Learn More
Medchemexpress LLC Trielaidin 25mg
SDP
Catalog No. 5000204624
Encompass_Preferred
TrielaidinIt is an organic compound belonging to the class of triacylglycerols It consists of three molecules of elaidic acid esterified to the glycerin backbone with a mild fatty smell TrieladinIt has a variety of applications in the cosmetic and personal care industries especially as an emollient and conditioning agent in skin and hair care products Additionally it is used as a lubricant and surfactant in industrial settings
Catalog No. 50-002-04624 Supplier Medchemexpress LLC Supplier No. 25MGHYW127506
TrielaidinIt is an organic compound belonging to the class of triacylglycerols It consists of three molecules of elaidic acid esterified to the glycerin backbone with a mild fatty smell TrieladinIt has a variety of applications in the cosmetic and personal care industries especially as an emollient and conditioning agent in skin and hair care products Additionally it is used as a lubricant and surfactant in industrial settings