Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trierucin, TCI America™
SDP

Catalog No. T13901G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T13901G Supplier TCI America Supplier No. T13901G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2752-99-0
Molecular Formula C69H128O6
Molecular Weight (g/mol) 1053.78
MDL Number MFCD00056321
InChI Key XDSPGKDYYRNYJI-IUPFWZBJSA-N
Synonym Glycerol Tri-13-docosenoate, 1,2,3-Tri(cis-13-docosenoyl) Glycerol
PubChem CID 5463075
IUPAC Name 1,3-bis[(13Z)-docos-13-enoyloxy]propan-2-yl (13Z)-docos-13-enoate
SMILES CCCCCCCC\C=C/CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC\C=C/CCCCCCCC)OC(=O)CCCCCCCCCCC\C=C/CCCCCCCC

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 1053.78
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Trierucin
RTECSNumber JR1239000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.