Learn More
Sigma Aldrich Fine Chemicals Biosciences Triethanolamine, 102-71-6, MFCD00002855, 100mL
Catalog No. 501656817
Encompass
Linear Formula (HOCH2CH2)3N, ≥99.0% (GC), Molecular Weight 149.19, bp 190-193 °C/5 mmHg (lit.), mp 17.9-21 °C (lit.), Synonym: 2,2′,2′′-Nitrilotriethanol, Tris(2-hydroxyethyl)amine
Catalog No. 50-165-6817 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 90279100ML
Linear Formula (HOCH2CH2)3N, ≥99.0% (GC), Molecular Weight 149.19, bp 190-193 °C/5 mmHg (lit.), mp 17.9-21 °C (lit.), Synonym: 2,2′,2′′-Nitrilotriethanol, Tris(2-hydroxyethyl)amine