Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethanolamine (2,2,2-Nitrilotriethanol), Reagents

Catalog No. 01336909
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass Bottle
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-336-909 500 mL Glass Bottle
01-336-911 4 L Poly Bottle
01-336-910 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-336-909 Supplier Reagents Holdings Llc Supplier No. C2342500500C
Add to Cart
Add to Cart

Triethanolamine (2,2,2-Nitrilotriethanol), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 102-71-6
Molecular Formula C6H15NO3
Molecular Weight (g/mol) 149.19
MDL Number MFCD00002855
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
Synonym 2,2,2-Nitrilotriethanol, Trolamine
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethan-1-ol
SMILES OCCN(CCO)CCO

Specifications

Boiling Range 335°C
Melting Point 21°C
Color Colorless to Slight Yellow
Boiling Point 335°C
Melting Range 21°C
Packaging Glass Bottle
Quantity 500 mL
Specific Gravity 1.1242
Grade Reagent
Physical Form Liquid
Chemical Name or Material Triethanolamine (2,2,2-Nitrilotriethanol)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.