Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethanolamine, 20% w/v, Reagents

Catalog No. 01333471
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-471 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-471 Supplier Reagents Holdings Llc Supplier No. CT001800500C
Add to Cart
Add to Cart

Triethanolamine, 20% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

CAS 7732-18-5, 102-71-6
Color Colorless to Pale Yellow
Boiling Point 335°C
Packaging Glass Bottle
Molecular Formula H2O
Quantity 500 mL
Specific Gravity 1.0267
Synonym 2,2,2-Nitrilotriethanol, Trolamine
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name water
Molecular Weight (g/mol) 18.02
Percent Purity 20%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Triethanolamine, 20% w/v
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.