Learn More
Boston Bioproducts Inc Triethanolamine Buffer (0.2 M, pH 8.0) - 250ml
Catalog No. NC2243846
Encompass
The Triethanolamine (TEA) Buffer is bifunctional solution that exhibits properties of both alcohols and amines. It is a zwitterionic buffer and can be used as a protein purification reagent in the pH range of 7-8.5 as well as a stationary phase for liquid chromatography. This buffer functions as neutralizing and emulsifying agent, allowing it to neutralize fatty acids and enhance the solubility of oils and other components that are not fully water-soluble.
This item is not returnable. View return policy
Catalog No. NC2243846 Supplier Boston Bioproducts Inc Supplier No. BB2382250ML
The Triethanolamine (TEA) Buffer is bifunctional solution that exhibits properties of both alcohols and amines. It is a zwitterionic buffer and can be used as a protein purification reagent in the pH range of 7-8.5 as well as a stationary phase for liquid chromatography. This buffer functions as neutralizing and emulsifying agent, allowing it to neutralize fatty acids and enhance the solubility of oils and other components that are not fully water-soluble.