Learn More
Medchemexpress LLC Triethanolamine borate | 283-56-7 | 99.68% | 25 G
SDP
Catalog No. 5000414469
Encompass_Preferred
Triethanolamine borate is a compound classified as a borate. It is frequently utilized in the production of personal care products and cosmetics, where it functions as an emulsifier, pH adjuster, and buffer. This compound aids in stabilizing and thickening products like lotions and creams, thereby enhancing their texture. Additionally, it finds application in industrial sectors for manufacturing lubricants and metalworking fluids. Triethanolamine borate also serves as a biochemical reagent for life science research.
  • Functions as an emulsifier, pH adjuster, and buffer
  • Helps stabilize and thicken products
  • Improves overall texture of lotions and creams
  • Used in industrial applications
  • Serves as a biochemical reagent for life science research
Catalog No. 50-004-14469 Supplier Medchemexpress LLC Supplier No. 25GHYD0902
Triethanolamine borate is a compound classified as a borate. It is frequently utilized in the production of personal care products and cosmetics, where it functions as an emulsifier, pH adjuster, and buffer. This compound aids in stabilizing and thickening products like lotions and creams, thereby enhancing their texture. Additionally, it finds application in industrial sectors for manufacturing lubricants and metalworking fluids. Triethanolamine borate also serves as a biochemical reagent for life science research.
  • Functions as an emulsifier, pH adjuster, and buffer
  • Helps stabilize and thicken products
  • Improves overall texture of lotions and creams
  • Used in industrial applications
  • Serves as a biochemical reagent for life science research