Learn More
Medchemexpress LLC Triethanolamine borate | 283-56-7 | 99.9% | 5 G
SDP
Catalog No. 5000409604
Encompass_Preferred
Triethanolamine borate is a compound classified as a borate. It finds common application in the production of personal care and cosmetic items, where it functions as an emulsifier, pH adjuster, and buffer. This substance assists in stabilizing and thickening products such as lotions and creams, thereby enhancing their overall texture. Additionally, it is utilized in industrial contexts for the manufacturing of lubricants and metalworking fluids.
  • Used as an emulsifier, pH adjuster, and buffer in personal care and cosmetics.
  • Helps stabilize and thicken lotions and creams, improving their texture.
  • Applied in industrial sectors for producing lubricants and metalworking fluids.
Catalog No. 50-004-09604 Supplier Medchemexpress LLC Supplier No. HYD09025G
Triethanolamine borate is a compound classified as a borate. It finds common application in the production of personal care and cosmetic items, where it functions as an emulsifier, pH adjuster, and buffer. This substance assists in stabilizing and thickening products such as lotions and creams, thereby enhancing their overall texture. Additionally, it is utilized in industrial contexts for the manufacturing of lubricants and metalworking fluids.
  • Used as an emulsifier, pH adjuster, and buffer in personal care and cosmetics.
  • Helps stabilize and thicken lotions and creams, improving their texture.
  • Applied in industrial sectors for producing lubricants and metalworking fluids.