Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethanolamine, Reagent, 99-107.4%, Spectrum™ Chemical
SDP

Catalog No. 18607565 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-565 200 L Amber Glass Bottle
18-607-561 6 x 500 mL Amber Glass Bottle
18-607-562 4 L Amber Glass Bottle
18-607-563 4 x 4 L Amber Glass Bottle
18-607-564 20 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-607-565 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T2564200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

T2564, 102-71-6, C6H15NO3

Spectrum™ Chemical Triethanolamine, Reagent is a is colorless organic compound that has a viscous texture and is both a tertiary amine and a triol. It is mainly used as an emulsifier and a surfactant. It neutralizes fatty acids while adjusting and buffering the pH. It is used in the fabrication of cosmetics and medicine such as ear drops, skin lotions, eye gels, shampoos and others. It also has a use in the laboratory as a complexing agent for aluminum ions in aqueous solutions. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 102-71-6
Molecular Formula C6H15NO3
Molecular Weight (g/mol) 149.19
MDL Number MFCD00002855
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethan-1-ol
SMILES OCCN(CCO)CCO

Specifications

CAS Max % 1
Boiling Point 335°C
Packaging Amber Glass Bottle
Quantity 200 L
Percent Purity 99.0 to 107.4%
Grade Reagent
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Triethanolamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.