Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethoxy-3-(2-imidazolin-1-yl)propylsilane

Catalog No. 5676050MLF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-760-50MLF 50 mL
56-760-10MLF 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 56-760-50MLF Supplier Sigma Aldrich Supplier No. 5676050MLF
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Additive for inorganic modification of block copolymersCatalyst for condensation reactionsModifier for organically modified layered silicates

Specifications

CAS 58068-97-6
Density 1.005 g/mL (at 20°C (literature))
Molecular Formula C12H26N2O3Si
Refractive Index n20/D 1.452
Linear Formula C12H26N2O3Si
MDL Number MFCD00042997
Quantity 50 mL
Synonym 1-(3-Triethoxysilylpropyl)-2-imidazoline; 3-(2-Imidazolin-1-yl)propyltriethoxysilane
Molecular Weight (g/mol) 274.43
Percent Purity ≥97.0% (NT)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.