Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethoxy(3-glycidyloxypropyl)silane 96.0+%, TCI America™
SDP

Catalog No. T267525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T267525G 25 g
T26755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T267525G Supplier TCI America Supplier No. T267525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2602-34-8
Molecular Formula C12H26O5Si
Molecular Weight (g/mol) 278.42
MDL Number MFCD01311705
InChI Key JXUKBNICSRJFAP-UHFFFAOYNA-N
Synonym Glycidyl 3-Triethoxysilylpropyl Ether
PubChem CID 102856
IUPAC Name triethoxy({3-[(oxiran-2-yl)methoxy]propyl})silane
SMILES CCO[Si](CCCOCC1CO1)(OCC)OCC

Specifications

Color Colorless
Boiling Point 135°C
Quantity 25 g
Formula Weight 278.42
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethoxy(3-glycidyloxypropyl)silane
EINECSNumber (2)-2071
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.