Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethoxymethylsilane

Catalog No. 339644500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-964-4500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 33-964-4500ML Supplier Sigma Aldrich Supplier No. 339644500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethoxymethylsilane

Specifications

CAS 88568-95-0
Melting Point 77°C to 80°C
Molecular Formula C13H18NO7P
Refractive Index n20/D 1.383
Linear Formula (CH3O)2P(O)CH(NHCO2CH2C6H5)CO2CH3
MDL Number MFCD00043304
Quantity 500 mL
Synonym Methyltriethoxysilane
Molecular Weight (g/mol) 331.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.