Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethoxyphenylsilane

Catalog No. 175609250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-560-9250G 250 g
17-560-91KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 17-560-9250G Supplier Sigma Aldrich Supplier No. 175609250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

A silicon-based nucleophile shown to be reactive in Pd-catalyzed cross-coupling reactions.

Specifications

CAS 780-69-8
Density 0.996 g/mL at 25°C
Boiling Point 112°C to 113°C (10 mmHg, lit.)
Molecular Formula C12H20O3Si
Refractive Index n20/D 1.461
Linear Formula C6H5Si(OC2H5)3
MDL Number MFCD00009065
Quantity 250 g
Synonym Phenyltriethoxysilane
Molecular Weight (g/mol) 240.37 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.