Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethoxysilane

Catalog No. 39014350ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-014-350ML 50 mL
39-014-310ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 39-014-350ML Supplier Sigma Aldrich Supplier No. 39014350ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Undergoes hydrosilylation reactions with olefins in the presence of Pt catalysts. Useful as a reducing agent.

Specifications

CAS 998-30-1
Density 0.89 g/mL at 25°C
Boiling Point 134°C to 135°C
Molecular Formula C6H16O3Si
Refractive Index n20/D 1.377
Linear Formula (C2H5O)3SiH
MDL Number MFCD00009061
Quantity 50 mL
Molecular Weight (g/mol) 164.27 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.