Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethoxysilane, 95%

Catalog No. AC164812500
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164812500 250 g Glass bottle
AC164810100 10 g Glass bottle
AC164810500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC164812500 Supplier Thermo Scientific Chemicals Supplier No. 164812500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 998-30-1 | C6H16O3Si | 164.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 998-30-1
Molecular Formula C6H16O3Si
Molecular Weight (g/mol) 164.28
MDL Number MFCD00009061
InChI Key PKDCQJMRWCHQOH-UHFFFAOYSA-N
Synonym triethoxysilane, silane, triethoxy, unii-8t460wdh89, c6h16o3si, si oet 3h;, c2h5o 3sih, 4-01-00-01359 beilstein handbook reference, triethoxysilane 10g, triethoxysilyl modified poly 1,2-butadiene, poly 1,2-butadiene , triethoxysilyl modified
PubChem CID 6327710
IUPAC Name triethoxysilicon
SMILES CCO[Si](OCC)OCC

Specifications

Melting Point -170.0°C
Color Colorless to Yellow
Density 0.8900g/mL
Boiling Point 134.0°C to 135.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.3762
Linear Formula (C2H5O)3SiH
Quantity 250 g
Beilstein 01,334
Fieser 11,554; 17,366
Specific Gravity 0.89
Solubility Information Solubility in water: reacts
Formula Weight 164.28
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Triethoxysilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Fatal if inhaled.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
EINECSNumber 213-650-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.