Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethyl 1,1,2-ethanetricarboxylate

Catalog No. T59854100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T59854100G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T59854100G Supplier Sigma Aldrich Supplier No. T59854100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethyl 1,1,2-ethanetricarboxylate

Specifications

CAS 7459-46-3
Density 1.074 g/mL (at 25°C (literature))
Boiling Point 99°C (0.5 mmHg)
Molecular Formula C11H18O6
Refractive Index n20/D 1.429 (literature)
Linear Formula C2H5O2CCH2CH(CO2C2H5)2
MDL Number MFCD00009154
Quantity 100 mg
Molecular Weight (g/mol) 246.26
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.